Thermo Scientific Chemicals

3-Fluoroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A14228.14
25 g, Each
Thermo Scientific Chemicals

3-Fluoroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A14228.14
25 g, Each
Quantity
Catalog number: A14228.14
also known as A14228-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
372-19-0
IUPAC Name
3-fluoroaniline
Molecular Formula
C6H6FN
InChI Key
QZVQQUVWFIZUBQ-UHFFFAOYSA-N
SMILES
NC1=CC=CC(F)=C1
Appearance (Color)
Clear colorless or yellow to brown
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text