Thermo Scientific Chemicals

Perfluoro(methylcyclohexane), 94%, Thermo Scientific Chemicals

Catalog number: A14233.22
100 g, Each
Thermo Scientific Chemicals

Perfluoro(methylcyclohexane), 94%, Thermo Scientific Chemicals

Catalog number: A14233.22
100 g, Each
Quantity
Catalog number: A14233.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
355-02-2
IUPAC Name
1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
Molecular Formula
C7F14
InChI Key
QIROQPWSJUXOJC-UHFFFAOYSA-N
SMILES
FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F
Identification (FTIR)
Conforms
Refractive Index
1.2755-1.2825 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥92.0%

Description

Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for fluorous biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for “fluorous” biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.

Solubility
Miscible with acetone and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text