Thermo Scientific Chemicals

4-Bromobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A14237.14
25 g, Each
Thermo Scientific Chemicals

4-Bromobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A14237.14
25 g, Each
Quantity
Catalog number: A14237.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1122-91-4
IUPAC Name
4-bromobenzaldehyde
Molecular Formula
C7H5BrO
InChI Key
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=O)C=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥98.0%
Free acid (titration)
≤1%
Identification (FTIR)
Conforms

Description

4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

Solubility
Soluble in ethanol and benzene. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text