4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals
4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14239.14
also known as A14239-14
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals
Catalog numberA14239.14
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS2785-89-9
IUPAC Name4-ethyl-2-methoxyphenol
Molecular FormulaC9H12O2
InChI KeyCHWNEIVBYREQRF-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)>97.5%
Refractive Index1.5260-1.5300 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only