Ethoxyacetylene, ca 50% w/w in hexanes, Thermo Scientific Chemicals
Ethoxyacetylene, ca 50% w/w in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethoxyacetylene, ca 50% w/w in hexanes, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A14245.14
also known as A14245-14
Price (USD)/ Each
352.65
Online exclusive
392.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
352.65
Online exclusive
392.00 
Save 39.35 (10%)
Add to cart
Ethoxyacetylene, ca 50% w/w in hexanes, Thermo Scientific Chemicals
Catalog numberA14245.14
Price (USD)/ Each
352.65
Online exclusive
392.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS927-80-0
IUPAC Nameethoxyethyne
Molecular FormulaC4H6O
InChI KeyWMYNMYVRWWCRPS-UHFFFAOYSA-N
SMILESCCOC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to red to dark brown
FormLiquid
Assay (GC)48.0-54.0% (w/w)
Refractive Index1.3750-1.3850 @ 20°C
Ethoxyacetylene is used in Arens-van Dorp synthesis to prepare propargyl alcohol via reaction with ketone. In organic synthesis, it undergoes [2+2] cycloaddition reactions with ketenes to give cyclobutenone derivatives. It is also used to prepare alpha, beta-unsaturated esters from ketones by following Meyer-Schuster rearrangement.Used with N-vinylamides and triflic anhydride in a direct, three-component synthesis of pyridines. Also used to prepare α, β-unsaturated esters from ketones via a Meyer-Schuster rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethoxyacetylene is used in Arens-van Dorp synthesis to prepare propargyl alcohol via reaction with ketone. In organic synthesis, it undergoes [2+2] cycloaddition reactions with ketenes to give cyclobutenone derivatives. It is also used to prepare alpha, beta-unsaturated esters from ketones by following Meyer-Schuster rearrangement.Used with N-vinylamides and triflic anhydride in a direct, three-component synthesis of pyridines. Also used to prepare α, β-unsaturated esters from ketones via a Meyer-Schuster rearrangement.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Supplied in solution to reduce polymerization. Quantities offered relate to gross weight of solution.
  2. Undergoes [2+2] cycloaddition reactions with ketenes to give cyclobutenone derivatives: J. Org. Chem., 38, 1451 (1973).
  3. Adds to carbonyl compounds in the presence of ZnBr2: Can. J. Chem., 54, 2310 (1976).
  4. The Grignard reagent, prepared by exchange with ethyl magnesium bromide, adds to carbonyl compounds to give carbinols which rearrange in acid to ɑß-unsaturated acids. Alternatively, partial reduction of the adduct followed by hydrolysis provides a route to ɑß-unsaturated aldehydes: J. Chem. Soc., 1823 (1949); Rec. Trav. Chim., 82, 305 (1963):
  5. Rearrangement by BF3 etherate leads directly to the ethyl ester: Rec. Trav. Chim., 78, 664 (1959); Chem. Lett., 1129 (1981); 241, 1271 (1982). See also Chloroacetaldehyde diethyl acetal, L07661.
  6. Has been used as a mild dehydrating agent in peptide synthesis: Rec. Trav. Chim., 77, 1153 (1958), and for conversion of sensitive dicarboxylic acids to their anhydrides: J. Org. Chem., 33, 3808 (1968).
  7. Aleman, G. H. J.; Machado, R. A. R.; Gorls, H.; Baldwin, I. T.; Bolan, W. Synthesis, structural characterization and biological activity of two diastereomeric JA-Ile macrolactones. Org. Biomol. Chem. 2015, 13 (21), 5885-5893.
  8. Henry, C.; Bolien, D.; Ibanescu, B.; Bloodworth, S.; Harrowven, D. C.; Zhang, X.; Craven, A. Generation and Trapping of Ketenes in Flow. Eur. J. Org. Chem. 2015, 2015 (7), 1491-1499.