Thermo Scientific Chemicals

Bathophenanthroline, 98+%, Thermo Scientific Chemicals

Catalog number: A14258.14
25 g, Each
Thermo Scientific Chemicals

Bathophenanthroline, 98+%, Thermo Scientific Chemicals

Catalog number: A14258.14
25 g, Each
Quantity
Catalog number: A14258.14
also known as A14258-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1662-01-7
IUPAC Name
4,7-diphenyl-1,10-phenanthroline
Molecular Formula
C24H16N2
InChI Key
DHDHJYNTEFLIHY-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1
Appearance (Color)
Pale cream to cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
217.0-224.0?C

Description

Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text