Thermo Scientific Chemicals

cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer, Thermo Scientific Chemicals

Catalog number: A14262.14
25 g, Each
Thermo Scientific Chemicals

cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer, Thermo Scientific Chemicals

Catalog number: A14262.14
25 g, Each
Quantity
Catalog number: A14262.14
also known as A14262-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1576-95-0
Assay (GC)
>96.0% (major isomer)
Refractive Index
1.4345-1.4395 @ 20?C
Appearance (Color)
Clear, colorless
Form
Liquid

Description

cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text