Thermo Scientific Chemicals

Phenylacetaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A14263.14
25 g, Each
Thermo Scientific Chemicals

Phenylacetaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A14263.14
25 g, Each
Quantity
Catalog number: A14263.14
also known as A14263-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-78-1
IUPAC Name
2-phenylacetaldehyde
Molecular Formula
C8H8O
InChI Key
DTUQWGWMVIHBKE-UHFFFAOYSA-N
SMILES
O=CCC1=CC=CC=C1
Refractive Index
1.5250-1.5350 @ 20°C
Free acid (titration)
≤4.5%
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms

Description

Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text