Thermo Scientific Chemicals

Benzoic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A14269.18
50 g, Each
Thermo Scientific Chemicals

Benzoic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A14269.18
50 g, Each
Quantity
Catalog number: A14269.18
also known as A14269-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-97-0
IUPAC Name
benzoyl benzoate
Molecular Formula
C14H10O3
InChI Key
CHIHQLCVLOXUJW-UHFFFAOYSA-N
SMILES
O=C(OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder or crystalline powder or crystals
Assay (chromatographic, GC or HPLC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
38-45°C

Description

Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.

Solubility
Soluble in alcohol, ether, chloroform, acetone, ethyl acetate, benzene, toluene, glacial acetic acid, acetic anhydride and ethanol. Slightly soluble in benzene and acetone. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text