Thermo Scientific Chemicals

1-Phenyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A14270.09
10 g, Each
Thermo Scientific Chemicals

1-Phenyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A14270.09
10 g, Each
Quantity
Catalog number: A14270.09
also known as A14270-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1126-00-7
Assay (GC)
≥97.5%
Appearance (Color)
Clear pale yellow to yellow
Refractive Index
1.5955-1.5995 @ 20?C
Form
Liquid

Description

1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text