Thermo Scientific Chemicals

1-Phenyl-1-propyne, 98%, Thermo Scientific Chemicals

Catalog number: A14272.06
5 g, Each
Thermo Scientific Chemicals

1-Phenyl-1-propyne, 98%, Thermo Scientific Chemicals

Catalog number: A14272.06
5 g, Each
Quantity
Catalog number: A14272.06
also known as A14272-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
673-32-5
IUPAC Name
(prop-1-yn-1-yl)benzene
Molecular Formula
C9H8
InChI Key
GHUURDQYRGVEHX-UHFFFAOYSA-N
SMILES
CC#CC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5620-1.5660 @ 20?C

Description

1-Phenyl-1-propyne is used as a intermediate for phrama industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-propyne is used as a intermediate for phrama industry and chemical research.

Solubility
Sparingly soluble in water (0.26 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text