Thermo Scientific Chemicals

3,4,5-Trimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A14274.22
100 g, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A14274.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14274.22
also known as A14274-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-41-2
IUPAC Name
3,4,5-trimethoxybenzoic acid
Molecular Formula
C10H12O5
InChI Key
SJSOFNCYXJUNBT-UHFFFAOYSA-N
SMILES
COC1=CC(=CC(OC)=C1OC)C(O)=O
Appearance (Color)
White to cream
Form
Crystalline powder
Assay (HPLC)
≥99.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
165-172?C

Description

3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text