Thermo Scientific Chemicals

2-Adamantanone, 98%, Thermo Scientific Chemicals

Catalog number: A14275.06
5 g, Each
Thermo Scientific Chemicals

2-Adamantanone, 98%, Thermo Scientific Chemicals

Catalog number: A14275.06
5 g, Each
Quantity
Catalog number: A14275.06
also known as A14275-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
700-58-3
IUPAC Name
adamantan-2-one
Molecular Formula
C10H14O
InChI Key
IYKFYARMMIESOX-UHFFFAOYSA-N
SMILES
O=C1C2CC3CC(C2)CC1C3
Form
Powder or lumps
Appearance (Color)
White
Assay (GC)
≥97.5%

Description

It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (±)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (+/-)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text