Thermo Scientific Chemicals

2-Amino-5-bromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14280.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14280.14
25 g, Each
Quantity
Catalog number: A14280.14
also known as A14280-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1072-97-5
IUPAC Name
5-bromopyridin-2-amine
Molecular Formula
C5H5BrN2
InChI Key
WGOLHUGPTDEKCF-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=N1
Assay (GC)
≥97.5%
Comment
May contain a small quantity of particulate matter from the manufacturing process
Identification (FTIR)
Conforms
Melting Point (clear melt)
135-145?C
Appearance (Color)
White to cream to pale brown

Description

2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text