2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.
Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
RUO – Research Use Only
Antonio Luque; Jon Sertucha; Luis Lezam; Teófilo Rojo; and Pascual Román. Synthesis, characterisation and crystal structure of 2-aminopyridinium(2-amino-5-bromopyridine)tribromocuprate(II)and bis(2-aminopyridinium)tetrabromocuprate(II). J. Chem. Soc., Dalton Trans. 1997, (5), 847-854.
N. Sundaraganesan; H. Saleem; S. Mohan. Vibrational spectra, assignments and normal coordinate analysis of 2-amino-5-bromopyridine. J. Chem. Soc., Dalton Trans. 2003, 59 (5), 1113-1118.