Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14288.06
5 g, Each
Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14288.06
5 g, Each
Quantity
Catalog number: A14288.06
also known as A14288-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5445-51-2
IUPAC Name
cyclobutane-1,1-dicarboxylic acid
Molecular Formula
C6H8O4
InChI Key
CCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILES
OC(=O)C1(CCC1)C(O)=O
Appearance (Color)
White to pale cream
Form
Powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text