1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals
1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14288.14
also known as A14288-14
Price (USD)/ Each
143.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
143.00
Add to cart
1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals
Catalog numberA14288.14
Price (USD)/ Each
143.00
-
Add to cart
Chemical Identifiers
CAS5445-51-2
IUPAC Namecyclobutane-1,1-dicarboxylic acid
Molecular FormulaC6H8O4
InChI KeyCCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILESOC(=O)C1(CCC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only