Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
Catalog number A14289.0I
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
Add to cart
Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Catalog numberA14289.0I
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS99-76-3
IUPAC Namemethyl 4-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyLXCFILQKKLGQFO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)124.0-130.0?C
Appearance (Color)White
FormPowder
Methyl 4-hydroxybenzoate is used as an anti-fungal agent. It is also used as a preservative in foods, beverages and cosmetics. It acts as an inhibitor of growth of molds and to lesser extent bacteria and as a vehicle for ophthalmic solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-hydroxybenzoate is used as an anti-fungal agent. It is also used as a preservative in foods, beverages and cosmetics. It acts as an inhibitor of growth of molds and to lesser extent bacteria and as a vehicle for ophthalmic solution.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Luo, H. Y.; Zhang, M.; Si, N. C.; Meng, M. J.; Yan, L.; Zhu, W. J.; Li, C. X. Molecularly imprinted open porous membranes made from Pickering W/O HIPEs for selective adsorption and separation of methyl 4-hydroxybenzoate. Chin. Chem. Lett. 2015, 26 (8), 1036-1041.
  2. Santos, I.; Ramos, C.; Ramos, J.; Oliveira, R.; Cunha, I. Efficient Association between PGF2α and Methyl 4-hydroxybenzoate Sex Pheromone Prior to Electroejaculation in Dogs. Reprod. Domest. Anim. 2013, 48 (1), 160-164.