Thermo Scientific Chemicals

Sodium 1-octanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A14292.06
5 g, Each
Thermo Scientific Chemicals

Sodium 1-octanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A14292.06
5 g, Each
Quantity
Catalog number: A14292.06
also known as A14292-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5324-84-5
IUPAC Name
sodium octane-1-sulfonate
Molecular Formula
C8H17NaO3S
InChI Key
HRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCS([O-])(=O)=O
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Powder
Water Content (Karl Fischer Titration)
≤1.5%
Assay (ex Sulphated Ash)
≥98.5%

Description

Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

Solubility
Soluble in water producing a clear and colorless solution

Notes
Sodium D-gluconate is stable under normal temperatures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text