Thermo Scientific Chemicals

2,6-Lutidine N-oxide, 98%, Thermo Scientific Chemicals

Catalog number: A14299.30
250 g, Each
Thermo Scientific Chemicals

2,6-Lutidine N-oxide, 98%, Thermo Scientific Chemicals

Catalog number: A14299.30
250 g, Each
Quantity
Catalog number: A14299.30
also known as A14299-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1073-23-0
IUPAC Name
2,6-dimethylpyridin-1-ium-1-olate
Molecular Formula
C7H9NO
InChI Key
LIDGFHXPUOJZMK-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=[N+]1[O-]
Form
Liquid
Refractive Index
1.5640-1.5720 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text