Thermo Scientific Chemicals

(Chloromethyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: A14303.18
50 g, Each
Thermo Scientific Chemicals

(Chloromethyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: A14303.18
50 g, Each
Quantity
Catalog number: A14303.18
also known as A14303-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2344-80-1
IUPAC Name
(chloromethyl)trimethylsilane
Molecular Formula
C4H11ClSi
InChI Key
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
SMILES
C[Si](C)(C)CCl
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4155-1.4195 @ 20?C
Assay (GC)
≥97.5%

Description

(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

Solubility
Soluble in water (0.4g/L at 20°C).

Notes
Store in cool, well ventilated area. Keep container tightly closed. Moisture sensitive. Store under argon.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text