Thermo Scientific Chemicals

Bispyrazolone, 98+%, Thermo Scientific Chemicals

Catalog number: A14308.06
5 g, Each
Thermo Scientific Chemicals

Bispyrazolone, 98+%, Thermo Scientific Chemicals

Catalog number: A14308.06
5 g, Each
Quantity
Catalog number: A14308.06
also known as A14308-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7477-67-0
IUPAC Name
5,5'-dimethyl-2,2'-diphenyl-2H,2'H,3H,3'H,4H,4'H-[4,4'-bipyrazole]-3,3'-dione
Molecular Formula
C20H18N4O2
InChI Key
FORCWSNQDMPPOC-UHFFFAOYNA-N
SMILES
CC1=NN(C(=O)C1C1C(C)=NN(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Elemental Analysis
N: 15.77-16.58% (Theory: 16.18%)
Elemental Analysis
C: 67.62-71.09% (Theory: 69.35%)
Appearance (Color)
White to cream or pale yellow
Assay (HPLC)
≥98.0%

Description

It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

Solubility
Solubility in hot DMF is almost transparent. Soluble in formic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text