Bispyrazolone, 98+%, Thermo Scientific Chemicals
Bispyrazolone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bispyrazolone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14308.14
also known as A14308-14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
Add to cart
Bispyrazolone, 98+%, Thermo Scientific Chemicals
Catalog numberA14308.14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7477-67-0
IUPAC Name5,5'-dimethyl-2,2'-diphenyl-2H,2'H,3H,3'H,4H,4'H-[4,4'-bipyrazole]-3,3'-dione
Molecular FormulaC20H18N4O2
InChI KeyFORCWSNQDMPPOC-UHFFFAOYNA-N
SMILESCC1=NN(C(=O)C1C1C(C)=NN(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Elemental AnalysisN: 15.77-16.58% (Theory: 16.18%)
Elemental AnalysisC: 67.62-71.09% (Theory: 69.35%)
Appearance (Color)White to cream or pale yellow
Assay (HPLC)≥98.0%
It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for the detection and determination of cyanide. It is also used in an experimental procedure, done in order to remove linamarin in starch from cassava roots, by using plant cell wall-degrading enzymes, xylanase and cellulase. It was also used for spectrophotometric determination and quantization of urea in urine samples. It is also used in assay of cyanogenic compounds and determination of ammonia.

Solubility
Solubility in hot DMF is almost transparent. Soluble in formic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Sharma V, et.al. Spectrophotometric Determination of Urea in Urine Samples by using Bispyrazolone Method.Proc. Indian Acad. Sci. A.,2013,79(1).
  2. Lidmila. Prochazkova.Spectrophotometric Determination of Ammonia as Rubazoic Acid with Bispyrazolone Reagent.Anal. Chem.,1964,36(4), 865-871.
  3. Reagent for the detection and determination of cyanide.