Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals
Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A14315.0E
also known as A14315-0E
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
Add to cart
Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals
Catalog numberA14315.0E
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
CommentK-vlaue 27-33
Water Content (Karl Fischer Titration)<5%
Appearance (Color)White to pale cream
FormPowder or flakes
Assay (unspecified)≥95% (dry weight basis)
View more
Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine. It is used as a capping agent to synthesize silver nanowires through a polyol process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only