Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals

Catalog number: A14315.22
100 g, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 58,000, Thermo Scientific Chemicals

Catalog number: A14315.22
100 g, Each
Quantity
Catalog number: A14315.22
also known as A14315-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-39-8
Molecular Formula
(C6H9NO)n
SMILES
*-CC(-*)N1CCCC1=O
MDL Number
MFCD01076626
Comment
K-vlaue 27-33
Water Content (Karl Fischer Titration)
<5%
Appearance (Color)
White to pale cream
Form
Powder or flakes
Assay (unspecified)
≥95% (dry weight basis)

Description

Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine. It is used as a capping agent to synthesize silver nanowires through a polyol process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text