3-Methoxyphenylacetonitrile, 99%, Thermo Scientific Chemicals
3-Methoxyphenylacetonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyphenylacetonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14317.14
also known as A14317-14
Price (USD)/ Each
124.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
124.00
Add to cart
3-Methoxyphenylacetonitrile, 99%, Thermo Scientific Chemicals
Catalog numberA14317.14
Price (USD)/ Each
124.00
-
Add to cart
Chemical Identifiers
CAS19924-43-7
IUPAC Name2-(3-methoxyphenyl)acetonitrile
Molecular FormulaC9H9NO
InChI KeyLXKNAUOWEJWGTE-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CC#N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to pale yellow
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5295-1.5325 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only