Thermo Scientific Chemicals

4-Bromo-2-methyl-6-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog number: A14319.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-methyl-6-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog number: A14319.14
25 g, Each
Quantity
Catalog number: A14319.14
also known as A14319-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77811-44-0
IUPAC Name
4-bromo-2-methyl-6-nitroaniline
Molecular Formula
C7H7BrN2O2
InChI Key
ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC(=C1N)[N+]([O-])=O
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point
141.5-147.5?C
Appearance (Color)
White to yellow or orange to brown

Description

It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

Solubility
Slightly soluble in water and soluble in hot methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text