Thermo Scientific Chemicals

N-Iodosuccinimide, 97%, Thermo Scientific Chemicals

Catalog number: A14320.22
100 g, Each
Thermo Scientific Chemicals

N-Iodosuccinimide, 97%, Thermo Scientific Chemicals

Catalog number: A14320.22
100 g, Each
Quantity
Catalog number: A14320.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
516-12-1
IUPAC Name
1-iodopyrrolidine-2,5-dione
Molecular Formula
C4H4INO2
InChI Key
LQZMLBORDGWNPD-UHFFFAOYSA-N
SMILES
IN1C(=O)CCC1=O
Form
Crystals or powder or crystalline powder.
Comment
Contains a small quantity of dark particulate matter from the manufacturing process.
Identification (FTIR)
Conforms
Appearance (Color)
White to yellow to brown
Assay (Iodometric Titration)
≥96.0 to ≤104.0%%

Description

N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

Solubility
Soluble in dioxane, tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text