Thermo Scientific Chemicals

Ethyl mercaptoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: A14321.22
100 g, Each
Thermo Scientific Chemicals

Ethyl mercaptoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: A14321.22
100 g, Each
Quantity
Catalog number: A14321.22
also known as A14321-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
623-51-8
IUPAC Name
ethyl 2-sulfanylacetate
Molecular Formula
C4H8O2S
InChI Key
PVBRSNZAOAJRKO-UHFFFAOYSA-N
SMILES
CCOC(=O)CS
Refractive Index
1.4555-1.4595 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Form
Liquid

Description

It finds its application in the reaction of the dilithio-derivative with an aldehyde, followed by conversion to the episulfide by reaction with ethyl chloroformate, and desulfurization with triethyl phosphite, leads to the (E)-2-alkenoic acid with high stereoselectivity. It is used as depilatories, hair straightening agent, hair waving agent, and reducing agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the reaction of the dilithio-derivative with an aldehyde, followed by conversion to the episulfide by reaction with ethyl chloroformate, and desulfurization with triethyl phosphite, leads to the (E)-2-alkenoic acid with high stereoselectivity. It is used as depilatories, hair straightening agent, hair waving agent, and reducing agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and store away from ignition sources. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text