Thermo Scientific Chemicals

2-Amino-5-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14329.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-5-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14329.14
25 g, Each
Quantity
Catalog number: A14329.14
also known as A14329-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
635-21-2
IUPAC Name
2-amino-5-chlorobenzoic acid
Molecular Formula
C7H6ClNO2
InChI Key
IFXKXCLVKQVVDI-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1C(O)=O
Appearance (Color)
Pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤1%
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps

Description

2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text