Thermo Scientific Chemicals

Allyl chloride, 98%, stab. with propylene oxide, Thermo Scientific Chemicals

Catalog number: A14330.AP
500 mL, Each
Thermo Scientific Chemicals

Allyl chloride, 98%, stab. with propylene oxide, Thermo Scientific Chemicals

Catalog number: A14330.AP
500 mL, Each
Quantity
Catalog number: A14330.AP
also known as A14330-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-05-1
IUPAC Name
3-chloroprop-1-ene
Molecular Formula
C3H5Cl
InChI Key
OSDWBNJEKMUWAV-UHFFFAOYSA-N
SMILES
ClCC=C
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4135-1.4175 @ 20?C
Appearance (Color)
Clear colourless
Assay (GC)
≥97.5%

Description

Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

Solubility
Soluble in water (3600 mg/L at 20°C), ethanol, ether, petroleum, acetone, chloroform, benzene, methanol and 2-prop.

Notes
Store at -20°C. Keep tightly closed in a dry and cool place. Keep in properly labeled containers. Keep containers tightly closed in a cool, well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents, boron trifluoride, sulfuric acid and nitric acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text