Thermo Scientific Chemicals

1-Bromoundecane, 98%, Thermo Scientific Chemicals

Catalog number: A14333.14
25 g, Each
Thermo Scientific Chemicals

1-Bromoundecane, 98%, Thermo Scientific Chemicals

Catalog number: A14333.14
25 g, Each
Quantity
Catalog number: A14333.14
also known as A14333-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-67-4
IUPAC Name
1-bromoundecane
Molecular Formula
C11H23Br
InChI Key
IKPSIIAXIDAQLG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCBr
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4550-1.4590 @ 20?C

Description

1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text