2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.
Solubility
Soluble in water (50g/L at 20°C)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Backx P H.; Gao W D.; Azan-Backx M D.; Marban E. Mechanism of force inhibition by 2,3-butanedione monoxime in rat cardiac muscle: roles of [Ca2+]i and cross-bridge kinetics. The Journal of Physiology. 1994, 476 (3), 487-500.
K. Horiuti, H. Higuchi.; Y. Umazume.; M. Konishi.; O. Okazaki.; S. Kurihara. Mechanism of action of 2, 3-butanedione 2-monoxime on contraction of frog skeletal muscle fibres. Journal of Muscle Research & Cell Motility. 1988, 9 (2), 156-164.
Reagent for colorimetric determination of urea: Anal. Biochem., 97, 421 (1979). Reagent for Co, Ni, Pd and Re: Talanta, 26, 425 (1979).
Formation of the silyl enol ether (1-aza-3-siloxy-1,3-butadiene), followed by hetero Diels-Alder reaction with Dimethyl acetylenedicarboxylate, A11437 affords a highly-functionalized pyridine: Tetrahedron, 62, 5454 (2006).