Thermo Scientific Chemicals

2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

Catalog number: A14343.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

Catalog number: A14343.06
5 g, Each
Quantity
Catalog number: A14343.06
also known as A14343-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
932-32-1
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow to yellow to brown
Assay (GC)
≥96.0%
Refractive Index
1.5780-1.5830 @ 20?C

Description

2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson's reagent and solvent tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson′s reagent and solvent tetrahydrofuran.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text