Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.
Solubility
Miscible with chloroform, alcohol, ether, dimethylsulfoxide, ethyl acetate and most organic solvents. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
- Sánchez, C. O.; Schott, E.; Zárate, X.; MacLeod-Carey, D.; Sobarzo, P.; Gatica, N. Effect of triphenylamine as electron-donor evenly spaced in 2, 4, 6 and 8 thiophene units of the main chain: synthesis and properties. Polym. Bull. 2015, 72 (4), 897-913.
- Sánchez, C. O.; Sobarzo, P.; Gatica, N. Electronic and structural properties of polymers based on phenylene vinylene and thiophene units. Control of the gap by gradual increases of thiophene moieties. New J. Chem. 2015, 39 (10), 7979-7987.