Thermo Scientific Chemicals

Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A14344.36
500 g, Each
Thermo Scientific Chemicals

Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A14344.36
500 g, Each
Quantity
Catalog number: A14344.36
also known as A14344-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-03-3
IUPAC Name
thiophene-2-carbaldehyde
Molecular Formula
C5H4OS
InChI Key
CNUDBTRUORMMPA-UHFFFAOYSA-N
SMILES
O=CC1=CC=CS1
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Free acid (titration)
<1.0%
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

Solubility
Miscible with chloroform, alcohol, ether, dimethylsulfoxide, ethyl acetate and most organic solvents. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text