Thermo Scientific Chemicals

4-Isopropoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14351.30
250 g, Each
Thermo Scientific Chemicals

4-Isopropoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14351.30
250 g, Each
Quantity
Catalog number: A14351.30
also known as A14351-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13205-46-4
IUPAC Name
4-(propan-2-yloxy)benzoic acid
Molecular Formula
C10H12O3
InChI Key
ZVERWTXKKWSSHH-UHFFFAOYSA-N
SMILES
CC(C)OC1=CC=C(C=C1)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
161.0-167.0?C

Description

4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text