Expi293™ Expression Medium
Interested in cGMP-banked Expi293F cells? Click HERE for product information.
Expi293™ Expression Medium
Gibco™

Expi293™ Expression Medium

Gibco Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidneyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A14351011 L
A143510310 L
A143510420 L
A14351026 x 1 L
Catalog number A1435101
Price (USD)
379.00
Each
-
Add to cart
Quantity:
1 L
Price (USD)
379.00
Each
Add to cart
Gibco Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.

• Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
• Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
• Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
• Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10L in disposable bioreactors
• Does not contain phenol red

Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

Product use
Customers using Expi293 Expression Medium in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell Line293E (HEK), 293 (HEK), Expi293F™, FreeStyle™ 293-F Cells, 293F
Cell TypeEmbryonic Kidney Cells
Concentration1 X
Culture EnvironmentCO2
Endotoxin Level< 1 EU/mL
Expression SystemExpi293F™
FormatBottle(s)
Included AntibioticsNo antibiotics
Inorganic SaltsCalcium, Magnesium, Sodium Phosphate
Osmolality275-295 mOsm/kg
Product LineExpi293™
Product TypeExpression Medium
Purity or Quality GradeCell Culture Grade
Quantity1 L
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesHigh Glucose, GlutaMAX, Sodium Pyruvate, Sodium Bicarbonate, Dextrose, Pyridoxine HCl
Without AdditivesNo HEPES, No Phenol Red, No Hypoxanthine, No Thymidine, No Insulin, No Vitamin A, No 2-Mercaptoethanol
Unit SizeEach
Contents & Storage
Store in the dark at +2°C to +8°C

Frequently asked questions (FAQs)

Can I purify my protein from Expi293 Expression Medium using ProBond or Ni-NTA purification systems?

Expi293 Expression Medium is not directly compatible with ProBond or Ni-NTA purification systems. We recommend performing a buffer exchange or dialyzing the samples before His-tag purification.

Can I use a different 293 cell line with the Expi293 Expression System?

Other 293 cell lines may be used with the Expi293 Expression System. However, before these cell lines may be used for transfection studies, they must be adapted to serum-free, suspension culture in Expi293 Expression Medium and evaluated for transfection and expression.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Which vector do you recommend using with the Expi293 Expression System?

We recommend using the pcDNA 3.4-TOPO TA vector (Cat. No. A14697). This vector contains the native, full-length CMV promoter and a WPRE (Woodchuck Posttranscriptional Regulatory Element) downstream of the cloning site, both of which contribute to high level gene expression (about 2-3 fold higher expression than with pcDNA3.3 TOPO vector, which in turn provides 2-5 fold higher expression than with a standard pcDNA vector). Of course, the expression level is also protein-dependent.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is the Expi293 Expression System completely animal-origin free (AOF)?

All the components of the system are animal-origin free except for the Opti-MEM I Reduced Serum Medium that is serum-free but not animal-origin free. Please see the Application Note for using the Expi293 Expression System under animal origin-free conditions:
http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.5943.file.dat/expi-293-animal-origin-free-co25751.pdf

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am interested in a mammalian expression system that will allow me to obtain very high yields of my protein. What kind of systems do you offer?

For stable high-yield expression, we offer the Freedom CHO-S Kit (Cat. No. A1369601) and Freedom DG44 Kit (Cat. No. A1373701). For transient high-yield expression, we offer the ExpiCHO Expression System (Cat. No. A29133), Expi293 Expression System (Cat. No. A14635), FreeStyle 293 Expression System (Cat. No. K900001), FreeStyle Max 293 Expression System (Cat. No. K900010), and FreeStyle Max CHO Expression System (Cat. No. K900020). For high-yield expression of functional membrane proteins in Exp293F cells, we offer the Expi293 MembranePro Expression System (Cat. Nos. A25869, A25870) that combines the scalability and ease of use of Expi293 and the technology of MembranePro to allow an increase of more than 20-fold in membrane protein yield compared to the standard, adherent culture MembranePro Functional Protein Expression System. Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/ProteinExpressionAnalysis/pdfs/PG1391-PJ6257-CO28704-Expi293Adaption-MembraneProTech_AppNoteGlobal-FHR.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link