Thermo Scientific Chemicals

Adamantane-1-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A14352.06
5 g, Each
Thermo Scientific Chemicals

Adamantane-1-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A14352.06
5 g, Each
Quantity
Catalog number: A14352.06
also known as A14352-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23074-42-2
IUPAC Name
adamantane-1-carbonitrile
Molecular Formula
C11H15N
InChI Key
FQFZASRJFRAEIH-UHFFFAOYSA-N
SMILES
N#CC12CC3CC(CC(C3)C1)C2
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
183-198?C
Appearance (Color)
White to pale cream or pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text