4,4'-Difluorobenzhydrol, 98+%, Thermo Scientific Chemicals
4,4'-Difluorobenzhydrol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Difluorobenzhydrol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14370.14
also known as A14370-14
Price (USD)/ Each
98.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
98.00
Add to cart
4,4'-Difluorobenzhydrol, 98+%, Thermo Scientific Chemicals
Catalog numberA14370.14
Price (USD)/ Each
98.00
-
Add to cart
Chemical Identifiers
CAS365-24-2
IUPAC Namebis(4-fluorophenyl)methanol
Molecular FormulaC13H10F2O
InChI KeyWCTZPQWLFWZYJE-UHFFFAOYSA-N
SMILESOC(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)43.0-50.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only