Thermo Scientific Chemicals

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Catalog number: A14376.30
250 g, Each
Thermo Scientific Chemicals

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Catalog number: A14376.30
250 g, Each
Quantity
Catalog number: A14376.30
also known as A14376-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-02-7
IUPAC Name
4-nitrophenol
Molecular Formula
C6H5NO3
InChI Key
BTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
White to cream to yellow to brown
Assay (GC)
≥98.5%
Melting Point (clear melt)
108-116?C
Form
Crystals, crystalline/granular powder and/or chunks
Identification (FTIR)
Conforms

Description

4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

Solubility
Freely soluble in alcohol, chloroform, ether. Freely soluble in acetone. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text