Thermo Scientific Chemicals

Neopentyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14390.14
25 g, Each
Thermo Scientific Chemicals

Neopentyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14390.14
25 g, Each
Quantity
Catalog number: A14390.14
also known as A14390-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-84-3
IUPAC Name
2,2-dimethylpropan-1-ol
Molecular Formula
C5H12O
InChI Key
KPSSIOMAKSHJJG-UHFFFAOYSA-N
SMILES
CC(C)(C)CO
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Appearance (Color)
Colorless to white
Melting Point (clear melt)
51.0-58.0°C
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid

Description

It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.

Solubility
Soluble in water at (20°C 40 g/L), alcohol, and ether. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text