Thermo Scientific Chemicals

2-Bromoterephthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14393.22
100 g, Each
Thermo Scientific Chemicals

2-Bromoterephthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14393.22
100 g, Each
Quantity
Catalog number: A14393.22
also known as A14393-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-35-6
IUPAC Name
2-bromobenzene-1,4-dicarboxylic acid
Molecular Formula
C8H5BrO4
InChI Key
QPBGNSFASPVGTP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C(O)=O)C(Br)=C1
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
295.0-304.0?C
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Powder or granules

Description

2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text