Thermo Scientific Chemicals

4-Hydroxycoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: A14394.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxycoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: A14394.22
100 g, Each
Quantity
Catalog number: A14394.22
also known as A14394-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1076-38-6
IUPAC Name
2-hydroxy-4H-chromen-4-one
Molecular Formula
C9H6O3
InChI Key
OWBBAPRUYLEWRR-UHFFFAOYSA-N
SMILES
OC1=CC(=O)C2=CC=CC=C2O1
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

Solubility
Soluble in ethanol and dimethyl formamide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text