4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number A14399.22
also known as A14399-22
Price (USD)/ Each
41.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
41.00
Add to cart
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA14399.22
Price (USD)/ Each
41.00
-
Add to cart
Chemical Identifiers
CAS104-01-8
IUPAC Name2-(4-methoxyphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeyNRPFNQUDKRYCNX-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream or pale yellow
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystalline powder and/or chunks
Melting Point (clear melt)84-88?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only