Thermo Scientific Chemicals

Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals

Catalog number: A14402.36
500 g, Each
Thermo Scientific Chemicals

Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals

Catalog number: A14402.36
500 g, Each
Quantity
Catalog number: A14402.36
also known as A14402-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-04-4
IUPAC Name
N,N,N-trimethylanilinium iodide
Molecular Formula
C9H14IN
InChI Key
KKLAORVGAKUOPZ-UHFFFAOYSA-M
SMILES
[I-].C[N+](C)(C)C1=CC=CC=C1
Assay (Titration ex Iodide)
≥98.5 to ≤101.5%
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder

Description

Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water and methanol.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text