Thermo Scientific Chemicals

Bromoacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14403.0B
1000 g, Each
Thermo Scientific Chemicals

Bromoacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14403.0B
1000 g, Each
Quantity
Catalog number: A14403.0B
also known as A14403-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-08-3
IUPAC Name
2-bromoacetic acid
Molecular Formula
C2H3BrO2
InChI Key
KDPAWGWELVVRCH-UHFFFAOYSA-N
SMILES
OC(=O)CBr
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder or fused solid
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
White to cream to yellow to pale brown
Assay (Silylated GC)
≥98.0%

Description

Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water, alcohol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to light and air. Handle and store under inert gas. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text