Thermo Scientific Chemicals

2,4,6-Trimethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A14407.18
50 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A14407.18
50 g, Each
Quantity
Catalog number: A14407.18
also known as A14407-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
938-18-1
IUPAC Name
2,4,6-trimethylbenzoyl chloride
Molecular Formula
C10H11ClO
InChI Key
UKRQMDIFLKHCRO-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C(Cl)=O)C(C)=C1
Appearance (Color)
Clear, colorless to yellow
Refractive Index
1.5270-1.5310 @ 20?C
Assay (Titration ex Chloride)
>98.0 to <102.0% (liberated Cl⁻ from reaction with Pyridine)
Form
Liquid
Assay (GC)
>98.0%

Description

2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

Solubility
Reacts slowly with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Air and moisture sensitive. Store under nitrogen. Incompatible with strong oxidizing agent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text