Ethoxymethylenemalononitrile, 97+%, Thermo Scientific Chemicals
Ethoxymethylenemalononitrile, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethoxymethylenemalononitrile, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14412.14
also known as A14412-14
Price (USD)/ Each
53.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
53.10
Add to cart
Ethoxymethylenemalononitrile, 97+%, Thermo Scientific Chemicals
Catalog numberA14412.14
Price (USD)/ Each
53.10
-
Add to cart
Chemical Identifiers
CAS123-06-8
IUPAC Name2-(ethoxymethylidene)propanedinitrile
Molecular FormulaC6H6N2O
InChI KeyOEICGMPRFOJHKO-UHFFFAOYSA-N
SMILESCCOC=C(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Cream to brown to yellow to orange
Melting Point61-68°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only