6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.
Solubility
Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.
Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
Reddy GN, et al. An improved synthesis of no-carrier-added (NCA) 6-[18 F] Fluoro-l-DOPA and its remote routine production for PET investigations of dopaminergic systems.Appl. Radiat. Isot.,1993,44(4), 645-49.
Butin AV, et al. o-Nitroaryl-bis (5-methylfur-2-yl) methanes as Versatile Synthons for the Synthesis of Nitrogen-Containing Heterocycles.Molecules.,1997,62(8), 62-8.